2-溴吡啶 ≥98.0% (通过GC)

供应商: TCI
Danger

B0650-100ML B0650-25ML B0650-500ML
TCIAB0650-100MLEA 0 CNY
TCIAB0650-100ML TCIAB0650-25ML TCIAB0650-500ML
2-溴吡啶 ≥98.0% (通过GC)
2-溴吡啶
分子式: C₅H₄BrN
沸点: 194°C
密度: 1,62 g/cm³ (20 °C)
闪点: 54°C
存储条件: Ambient
MDL编号: MFCD00006219
CAS号: 109-04-6
EINECS: 203-641-6
UN号: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4325252c307a8e2a91e4b2452010866a7fba0556&vid=4325252c307a8e2a91e4b2452010866a7fba0556&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR