2,3-二溴吡啶 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
D2917-1G
D2917-5G
TCIAD2917-1GEA
0
CNY
TCIAD2917-1G
TCIAD2917-5G
2,3-二溴吡啶 ≥98.0% (by GC)
2,3-二溴吡啶
分子式:
C₅H₃Br₂N 沸点: 249°C 溶点: 58…59 °C |
MDL编号:
MFCD00234014 CAS号: 13534-89-9 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...