2-(溴甲基)萘 ≥97.0% (by GC)
供应商: TCI
|
Danger
|
B1524-25G
B1524-5G
TCIAB1524-25GEA
0
CNY
TCIAB1524-25G
TCIAB1524-5G
2-(溴甲基)萘 ≥97.0% (by GC)
2-(溴甲基)萘
分子式:
C₁₁H₉Br 沸点: 214°C 溶点: 60…62 °C 存储条件: Refrigerator |
MDL编号:
MFCD00004123 CAS号: 939-26-4 EINECS: 213-359-5 UN号: 3261 ADR: 8,II |
|
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...