1-萘酚 ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: 1-羟基萘, α-萘酚

N0026-25G N0026-500G
TCIAN0026-25GEA 0 CNY
TCIAN0026-25G TCIAN0026-500G
1-萘酚 ≥99.0% (by GC)
1-萘酚
分子式: C₁₀H₈O
分子量: 144.17 g/mol
沸点: 147°C
溶点: 94
闪点: 125
存储条件: Ambient
MDL编号: MFCD00003930
CAS号: 90-15-3
EINECS: 201-969-4
UN号: 2811
ADR: 6.1 III
Merck Index: 14,06383
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b6956a5f9061b71cbb331e826794092518aa20&vid=12b6956a5f9061b71cbb331e826794092518aa20&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 99.0 %
Melting point 95.0 to 97.0 deg-C
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR