2,6-萘二羧酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2,6-Naphthalenedicarboxylic Acid, Naphthalene-2,6-dicarboxylic acid
N0377-100G
N0377-25G
TCIAN0377-100GEA
0
CNY
TCIAN0377-100G
TCIAN0377-25G
2,6-萘二羧酸 ≥98.0% (by GC, titration analysis)
2,6-萘二羧酸
分子式:
C₁₂H₈O₄ 分子量: 216.19 g/mol 沸点: 422 |
MDL编号:
MFCD00004105 CAS号: 1141-38-4 EINECS: 214-527-0 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Pyridine | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...