Ethyl 2-chloronicotinate ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

E0844-100G E0844-25G
TCIAE0844-100GEA 0 CNY
TCIAE0844-100G TCIAE0844-25G
Ethyl 2-chloronicotinate ≥98.0% (by GC)
Ethyl-2-chloronicotinate
分子式: C₈H₈ClNO₂
分子量: 185.61 g/mol
MDL编号: MFCD00674129
CAS号: 1452-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae69b6cfb9545b401121f95d35f14cd7c98de76b&vid=ae69b6cfb9545b401121f95d35f14cd7c98de76b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR