二苯基碘鎓六氟磷酸盐 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

D2238-1G D2238-25G D2238-5G
TCIAD2238-1GEA 0 CNY
TCIAD2238-1G TCIAD2238-25G TCIAD2238-5G
二苯基碘鎓六氟磷酸盐 ≥97.0% (by titrimetric analysis)
二苯基碘鎓六氟磷酸盐
分子式: C₁₂H₁₀F₆IP
MDL编号: MFCD00061398
CAS号: 58109-40-3
EINECS: 261-134-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0025593916b98ef863cd4a206ef5c1ff74958a7a&vid=0025593916b98ef863cd4a206ef5c1ff74958a7a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 97.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR