二苯基碘酰氯 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
D2356-25G
D2356-5G
TCIAD2356-25GEA
0
CNY
TCIAD2356-25G
TCIAD2356-5G
二苯基碘酰氯 ≥98.0% (by titrimetric analysis)
二苯基碘酰氯
分子式:
C₁₂H₁₀ClI 分子量: 316.57 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00011909 CAS号: 1483-72-3 EINECS: 216-049-8 UN号: 2811 ADR: 6.1 III |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...