Aminoguanidine hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride

A1129-25G A1129-500G
TCIAA1129-25GEA 0 CNY
TCIAA1129-25G TCIAA1129-500G
Aminoguanidine hydrochloride ≥98.0% (by titrimetric analysis)
Aminoguanidine hydrochloride
分子式: CH₇ClN₄
分子量: 110.55 g/mol
MDL编号: MFCD00039074
CAS号: 1937-19-5
EINECS: 217-707-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e0b3117e3d2e3ddc5fe16b0e86bac10b21f927a&vid=0e0b3117e3d2e3ddc5fe16b0e86bac10b21f927a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Melting point 161.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR