丁炔二酸二甲酯 ≥96.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Dimethyl 2-butynedioate
A0090-25ML
A0090-500ML
TCIAA0090-25MLEA
0
CNY
TCIAA0090-25ML
TCIAA0090-500ML
丁炔二酸二甲酯 ≥96.0% (by GC)
丁炔二酸二甲酯
分子式:
C₆H₆O₄ 沸点: 198°C 闪点: 86°C 存储条件: Refrigerator |
MDL编号:
MFCD00008456 CAS号: 762-42-5 EINECS: 212-098-4 UN号: 1760 ADR: 8,III |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...