四乙二醇二甲醚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
二甲基四甘醇, 双[2-(2-甲氧基乙氧基)乙基]醚, 四甘醇二甲醚, 2,5,8,11,14-五氧杂十五烷, 二甲氧基四乙二醇
B0497-25ML
B0497-500ML
TCIAB0497-25MLEA
0
CNY
TCIAB0497-25ML
TCIAB0497-500ML
四乙二醇二甲醚 ≥98.0% (by GC)
四乙二醇二甲醚
分子式:
C₁₀H₂₂O₅ 分子量: 222.28 g/mol 沸点: 276°C 闪点: 140°C |
MDL编号:
MFCD00008505 CAS号: 143-24-8 EINECS: 205-594-7 Merck Index: 14,09208 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...