3,4-二氢-2H-吡喃 ≥97.0% (by GC)

供应商: TCI
Danger

D0555-25ML D0555-500ML
TCIAD0555-25MLEA 0 CNY
TCIAD0555-25ML TCIAD0555-500ML
3,4-二氢-2H-吡喃 ≥97.0% (by GC)
3,4-二氢-2H-吡喃
分子式: C₅H₈O
分子量: 84.12 g/mol
沸点: 86°C
闪点: –6°C
MDL编号: MFCD00006558
CAS号: 110-87-2
EINECS: 203-810-4
UN号: 2376
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce63b35d82bafde9ec26e24d77119ceafa069ecc&vid=ce63b35d82bafde9ec26e24d77119ceafa069ecc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR