N,N′-二叔丁基乙二胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 1,2-双(叔丁基氨基)乙烷

D2638-25ML D2638-500ML
TCIAD2638-25MLEA 0 CNY
TCIAD2638-25ML TCIAD2638-500ML
N,N′-二叔丁基乙二胺 ≥98.0% (by GC, titration analysis)
N,N′-二叔丁基乙二胺
分子式: C₁₀H₂₄N₂
沸点: 198°C
闪点: 62°C
MDL编号: MFCD00014996
CAS号: 4062-60-6
EINECS: 223-769-6
UN号: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2edad7331c494dc566c447687a764d8db5a2ef76&vid=2edad7331c494dc566c447687a764d8db5a2ef76&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR