吡啶甲酸甲酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Methyl pyridine-2-carboxylate
P0423-25G
P0423-250G
TCIAP0423-25GEA
0
CNY
TCIAP0423-25G
TCIAP0423-250G
吡啶甲酸甲酯 ≥98.0% (by GC)
吡啶甲酸甲酯
分子式:
C₇H₇NO₂ 分子量: 137.14 g/mol 沸点: 233°C 闪点: 103°C |
MDL编号:
MFCD00038038 CAS号: 2459-07-6 EINECS: 219-545-2 |
|
产品技术规格
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...