2-溴吡啶-4-甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-溴异烟酸
B3368-1G
B3368-5G
TCIAB3368-1GEA
0
CNY
TCIAB3368-1G
TCIAB3368-5G
2-溴吡啶-4-甲酸 ≥98.0% (by GC, titration analysis)
2-溴吡啶-4-甲酸
分子式:
C₆H₄BrNO₂ 分子量: 202.01 g/mol 溶点: 229°C |
MDL编号:
MFCD01646069 CAS号: 66572-56-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...