Bis(2-ethylhexyl)azelate ≥75.0% (通过GC)

供应商: TCI

A0653-25ML A0653-500ML
TCIAA0653-25MLEA 0 CNY
TCIAA0653-25ML TCIAA0653-500ML
Bis(2-ethylhexyl)azelate ≥75.0% (通过GC)
Bis(2-ethylhexyl)azelate
分子式: C₂₅H₄₈O₄
闪点: 211°C
MDL编号: MFCD00040496
CAS号: 103-24-2
EINECS: 203-091-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d52f9da20bccc0cef3a7d70dc0e13e17065d2c8&vid=6d52f9da20bccc0cef3a7d70dc0e13e17065d2c8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 75.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR