反-1,4-二甲基环己烷 ≥95.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
反六氢对二甲苯
D0271-25ML
D0271-5ML
TCIAD0271-25MLEA
0
CNY
TCIAD0271-25ML
TCIAD0271-5ML
反-1,4-二甲基环己烷 ≥95.0% (by GC)
反-1,4-二甲基环己烷
分子式:
C₈H₁₆ 分子量: 112.22 g/mol 沸点: 119°C 溶点: -37 密度: 0.763 闪点: 6°C 存储条件: Ambient |
MDL编号:
MFCD00064955 CAS号: 2207-04-7 EINECS: 218-622-8 UN号: 2263 ADR: 3 II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...