4-tert-Butylcyclohexylamine cis- and trans-mixture ≥98.0% (通过滴定分析)

供应商: TCI
Danger

B1084-25ML B1084-250ML
TCIAB1084-25MLEA 0 CNY
TCIAB1084-25ML TCIAB1084-250ML
4-tert-Butylcyclohexylamine cis- and trans-mixture ≥98.0% (通过滴定分析)
4-tert-Butylcyclohexylamine
分子式: C₁₀H₂₁N
沸点: 202 °C (1013 hPa)
溶点: 18 °C
密度: 0,87 g/cm³ (20 °C)
闪点: 79°C
MDL编号: MFCD00013763
CAS号: 5400-88-4
EINECS: 226-444-7
UN号: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a870c61c6d6460f70b51628114acbf1d1246eb25&vid=ab52a0ea818ab17473b04126fbec455ce57a9ad7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR