4-甲基苯酐 ≥98.0% (通过GC,滴定分析)
供应商: TCI
|
Warning
|
M0559-25G
M0559-500G
TCIAM0559-25GEA
0
CNY
TCIAM0559-25G
TCIAM0559-500G
4-甲基苯酐 ≥98.0% (通过GC,滴定分析)
4-甲基苯酐
分子式:
C₉H₆O₃ 沸点: 299°C |
MDL编号:
MFCD00041856 CAS号: 19438-61-0 EINECS: 243-073-6 |
|
产品技术规格
Appearance | White to Yellow to Orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Morpholine Method) | min. 98.0 % |
Melting point | 89.0 to 94.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...