Methyl-2-bromo-5-chlorobenzoate ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Methyl 2-bromo-5-chlorobenzoate
M2033-25G
M2033-5G
TCIAM2033-25GEA
0
CNY
TCIAM2033-25G
TCIAM2033-5G
Methyl-2-bromo-5-chlorobenzoate ≥98.0% (by GC)
Methyl-2-bromo-5-chlorobenzoate
分子式:
C₈H₆BrClO₂ 分子量: 249.49 g/mol |
MDL编号:
MFCD00144763 CAS号: 27007-53-0 |
|
产品技术规格
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...