2-溴-1-乙基吡啶四氟硼酸盐 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: BEP

B1036-25G B1036-5G
TCIAB1036-25GEA 0 CNY
TCIAB1036-25G TCIAB1036-5G
2-溴-1-乙基吡啶四氟硼酸盐 ≥98.0% (by HPLC, titration analysis)
2-溴-1-乙基吡啶四氟硼酸盐
分子式: C₇H₉BBrF₄N
分子量: 273.86 g/mol
MDL编号: MFCD00060046
CAS号: 878-23-9
EINECS: 212-900-2
UN号: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=994a9cf09456337ec31e28c0a4f45aa4e9b01804&vid=994a9cf09456337ec31e28c0a4f45aa4e9b01804&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 103.0 to 106.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR