四丁基三氟甲磺酸铵 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
三氟甲磺酸, 四正丁铵三氟甲磺酸盐
T1568-10G
T1568-25G
TCIAT1568-10GEA
0
CNY
TCIAT1568-10G
TCIAT1568-25G
四丁基三氟甲磺酸铵 ≥98.0% (by titrimetric analysis)
四丁基三氟甲磺酸铵
分子式:
C₁₇H₃₆F₃NO₃S 分子量: 391.54 g/mol |
MDL编号:
MFCD00042585 CAS号: 35895-70-6 EINECS: 252-783-5 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 113.0 to 115.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...