3-Picolyl chloride hydrochloride ≥98.0% (通过HPLC,滴定分析)

供应商: TCI
Danger

C0825-100G C0825-25G
TCIAC0825-100GEA 0 CNY
TCIAC0825-100G TCIAC0825-25G
3-Picolyl chloride hydrochloride ≥98.0% (通过HPLC,滴定分析)
3-Picolyl chloride hydrochloride
分子式: C₆H₇Cl₂N
MDL编号: MFCD00012818
CAS号: 6959-48-4
EINECS: 230-150-4
UN号: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b0d23395ce85b1193238ebbaf3f5444b7669339&vid=3b0d23395ce85b1193238ebbaf3f5444b7669339&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR