4-乙炔基甲苯 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-甲苯基乙炔

E0655-25G E0655-5G
TCIAE0655-25GEA 0 CNY
TCIAE0655-25G TCIAE0655-5G
4-乙炔基甲苯 ≥97.0% (by GC)
4-乙炔基甲苯
分子式: C₉H₈
分子量: 116.16 g/mol
沸点: 168°C
闪点: 48°C
MDL编号: MFCD00008571
CAS号: 766-97-2
UN号: 1325
ADR: 4.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1485a049b4d392b2b4b369bec7949672b97444&vid=bf1485a049b4d392b2b4b369bec7949672b97444&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR