Ethyl 2,5-dichlorobenzoate ≥98.0% (by GC)
供应商: TCI
Synonyms:
Ethyl 2,5-dichlorobenzoate
D2566-25G
D2566-5G
TCIAD2566-25GEA
0
CNY
TCIAD2566-25G
TCIAD2566-5G
Ethyl 2,5-dichlorobenzoate ≥98.0% (by GC)
Ethyl-2,5-dichlorobenzoate
分子式:
C₉H₈Cl₂O₂ |
MDL编号:
MFCD00013636 CAS号: 35112-27-7 EINECS: 252-373-6 |
|
产品技术规格
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...