三乙醇铝 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 三乙氧基铝

A0245-25G A0245-250G
TCIAA0245-25GEA 0 CNY
TCIAA0245-25G TCIAA0245-250G
三乙醇铝 ≥98.0% (by titrimetric analysis)
三乙醇铝
分子式: C₆H₁₅AlO₃
沸点: 200°C
溶点: 140°C
MDL编号: MFCD00009658
CAS号: 555-75-9
EINECS: 209-105-8
UN号: 2925
ADR: 4.1,II
Merck Index: 14,00338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e3da3d19032ee96c2a013089d94bbbeecf9e81&vid=26e3da3d19032ee96c2a013089d94bbbeecf9e81&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR