环己烷 ≥99.5% (by GC)

供应商: TCI
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

C0469-25ML C0469-500ML
TCIAC0469-25MLEA 0 CNY
TCIAC0469-25ML TCIAC0469-500ML
环己烷 ≥99.5% (by GC)
环己烷
分子式: C₆H₁₂
沸点: 81°C
溶点: 7 °C
密度: 0,7785 g/cm³ (20 °C)
闪点: –20°C
存储条件: Ambient
MDL编号: MFCD00003814
CAS号: 110-82-7
EINECS: 203-806-2
UN号: 1145
ADR: 3,II
Merck Index: 14,02723
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b07639659b2a13e27fbc013757200f70c5b4807b&vid=b07639659b2a13e27fbc013757200f70c5b4807b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 99.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR