硝基环己烷 ≥95.0% (by GC)

供应商: TCI
Danger

N0187-25G N0187-5G
TCIAN0187-25GEA 0 CNY
TCIAN0187-25G TCIAN0187-5G
硝基环己烷 ≥95.0% (by GC)
硝基环己烷
分子式: C₆H₁₁NO₂
沸点: 113°C
密度: 1,0853 g/cm³ (0 °C)
闪点: 74°C
MDL编号: MFCD00003843
CAS号: 1122-60-7
EINECS: 214-354-0
UN号: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a7557aabf8afe1a667854680fe6be6494fa3eeb&vid=7a7557aabf8afe1a667854680fe6be6494fa3eeb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR