环丁基氯 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
氯环丁烷
C1033-1ML
C1033-5ML
TCIAC1033-1MLEA
0
CNY
TCIAC1033-1ML
TCIAC1033-5ML
环丁基氯 ≥98.0% (by GC)
环丁基氯
分子式:
C₄H₇Cl 分子量: 90.55 g/mol 沸点: 82°C 密度: 0.991 闪点: –8°C 存储条件: Refrigerator |
MDL编号:
MFCD00013736 CAS号: 1120-57-6 EINECS: 214-314-2 UN号: 1993 ADR: 3 II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...