8-羟基喹啉 ≥99.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
8-氢氧化喹啉, 8-羟基氮杂萘, 喔星
H0305-25G
H0305-500G
TCIAH0305-25GEA
0
CNY
TCIAH0305-25G
TCIAH0305-500G
8-羟基喹啉 ≥99.0% (by GC, titration analysis)
8-羟基喹啉
分子式:
C₉H₇NO 分子量: 145.16 g/mol 沸点: 126°C 溶点: 71.2 存储条件: Ambient |
MDL编号:
MFCD00006807 CAS号: 148-24-3 EINECS: 205-711-1 UN号: 3077 ADR: 9 III Merck Index: 14,04843 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 72.0 to 76.0 deg-C |
Solubility in Dichloromethane | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...