2-羟基喹啉 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 喹诺酮, 2-喹啉醇

H0304-1G H0304-10G
TCIAH0304-1GEA 0 CNY
TCIAH0304-1G TCIAH0304-10G
2-羟基喹啉 ≥98.0% (by GC)
2-羟基喹啉
分子式: C₉H₇NO
分子量: 145.16 g/mol
沸点: 347
存储条件: Ambient
MDL编号: MFCD00006743
CAS号: 59-31-4
EINECS: 200-420-6
Merck Index: 14,01825
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=680efe8f2dbf03e8dd0075c70ab71b589f1e668c&vid=bbfc697e64510df892015b19092822fbff00a86a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 194.0 to 201.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR