6-氯喹啉 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

C0283-1G C0283-5G
TCIAC0283-1GEA 0 CNY
TCIAC0283-1G TCIAC0283-5G
6-氯喹啉 ≥98.0% (by GC, titration analysis)
6-氯喹啉
分子式: C₉H₆ClN
分子量: 163.61 g/mol
沸点: 264°C
溶点: 43
存储条件: Refrigerator
MDL编号: MFCD00024024
CAS号: 612-57-7
EINECS: 210-314-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f6234f00615c9a15743ddddcfb9577aca9059d&vid=cb8c5c636a9d57a818890544bb02c48ff6075e13&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR