1,4-Dicyclohexylbenzene ≥98.0% (by GC)
供应商: TCI
D1028-25G
D1028-250G
TCIAD1028-25GEA
0
CNY
TCIAD1028-25G
TCIAD1028-250G
1,4-Dicyclohexylbenzene ≥98.0% (by GC)
1,4-Dicyclohexylbenzene
分子式:
C₁₈H₂₆ 沸点: 195°C 密度: 1,049 g/cm³ (0 °C) |
MDL编号:
MFCD00001452 CAS号: 1087-02-1 EINECS: 214-121-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...