1-Bromo-4-cyclohexylbenzene ≥97.0% (by GC)

供应商: TCI

B2686-25G B2686-5G
TCIAB2686-25GEA 0 CNY
TCIAB2686-25G TCIAB2686-5G
1-Bromo-4-cyclohexylbenzene ≥97.0% (by GC)
1-Bromo-4-cyclohexylbenzene
分子式: C₁₂H₁₅Br
分子量: 239.16 g/mol
MDL编号: MFCD00019339
CAS号: 25109-28-8
EINECS: 246-623-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0218719730e4f43c2feb66b70878254e6c368d0&vid=508efaf9632d4a97586da5d05fdcf93a0f2da3d1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR