1,2,3,5-Tetramethylbenzene ≥70% (通过GC)

供应商: TCI

T0565-25ML T0565-5ML
TCIAT0565-25MLEA 0 CNY
TCIAT0565-25ML TCIAT0565-5ML
1,2,3,5-Tetramethylbenzene ≥70% (通过GC)
1,2,3,5-Tetramethylbenzene

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 70.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR