1,3,5-三异丙基苯 ≥95.0% (by GC)

供应商: TCI
Warning

T0458-25ML T0458-500ML
TCIAT0458-25MLEA 0 CNY
TCIAT0458-25ML TCIAT0458-500ML
1,3,5-三异丙基苯 ≥95.0% (by GC)
1,3,5-三异丙基苯
分子式: C₁₅H₂₄
分子量: 204.36 g/mol
沸点: 235°C
溶点: -7
闪点: 86°C
存储条件: Ambient
MDL编号: MFCD00008890
CAS号: 717-74-8
EINECS: 211-941-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f382e17a04a2858d9f799ddadd3a44cbd44dae58&vid=7c6d10a56428570697500eea6c693e4fe61b4ab4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR