3-乙基甲苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3-Ethyltoluene, 1-Ethyl-3-methylbenzene
E0185-25ML
E0185-5ML
TCIAE0185-25MLEA
0
CNY
TCIAE0185-25ML
TCIAE0185-5ML
3-乙基甲苯 ≥98.0% (by GC)
3-乙基甲苯
分子式:
C₉H₁₂ 沸点: 160°C 密度: 0,869 g/cm³ (20 °C) 闪点: 38°C 存储条件: Ambient |
MDL编号:
MFCD00009259 CAS号: 620-14-4 EINECS: 210-626-8 UN号: 3295 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...