1,3-二异丙基苯 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
间二异丙苯
D2490-25ML
D2490-500ML
TCIAD2490-25MLEA
0
CNY
TCIAD2490-25ML
TCIAD2490-500ML
1,3-二异丙基苯 ≥96.0% (by GC)
1,3-二异丙基苯
分子式:
C₁₂H₁₈ 沸点: 204°C 溶点: –40 °C 密度: 0,85593 g/cm³ (20 °C) 闪点: 76°C 存储条件: Ambient |
MDL编号:
MFCD00008889 CAS号: 99-62-7 EINECS: 202-773-1 UN号: 3082 ADR: 9,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |