2,6-二叔丁基苯酚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1,3-Di(dimethylethyl)-2-hydroxybenzene
D0941-25G
D0941-500G
TCIAD0941-25GEA
0
CNY
TCIAD0941-25G
TCIAD0941-500G
2,6-二叔丁基苯酚 ≥98.0% (by GC)
2,6-二叔丁基苯酚
分子式:
C₁₄H₂₂O 沸点: 253°C 溶点: 34…37 °C 密度: 0,914 g/cm³ (20 °C) 闪点: 107 °C 存储条件: Ambient |
MDL编号:
MFCD00008820 CAS号: 128-39-2 EINECS: 204-884-0 ADR: 9,III |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 35.0 to 37.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...