2,6-二异丙基苯胺 ≥90.0% (通过GC)

供应商: TCI
Warning

D1755-25ML D1755-500ML
TCIAD1755-25MLEA 0 CNY
TCIAD1755-25ML TCIAD1755-500ML
2,6-二异丙基苯胺 ≥90.0% (通过GC)
2,6-二异丙基苯胺
分子式: C₁₂H₁₉N
沸点: 122°C
溶点: -45°C
密度: 0,94 g/cm³ (20 °C)
闪点: 117°C
存储条件: Ambient
MDL编号: MFCD00008887
CAS号: 24544-04-5
EINECS: 246-305-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=833278fe351a9cc9b41ca690194bd446a6eff255&vid=a4b56f3c6ab126c4d4c50351bc7eac206b1e5206&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR