2-氟-1,3-二甲苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-氟间二甲苯

F0402-25G F0402-5G
TCIAF0402-25GEA 0 CNY
TCIAF0402-25G TCIAF0402-5G
2-氟-1,3-二甲苯 ≥98.0% (by GC)
2-氟-1,3-二甲苯
分子式: C₈H₉F
分子量: 124.16 g/mol
沸点: 167°C
密度: 0.988
闪点: 31°C
MDL编号: MFCD00039215
CAS号: 443-88-9
EINECS: 207-144-5
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a583688b9256a147878a072ede2fdd5db8eefaab&vid=550149ed6eed4c3eb0a018c82a2e7fcbfaad793c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR