4-氯-1,2-二甲苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-氯邻二甲苯, 3,4-二甲基氯苯
C0356-100G
C0356-25G
TCIAC0356-100GEA
0
CNY
TCIAC0356-100G
TCIAC0356-25G
4-氯-1,2-二甲苯 ≥98.0% (by GC)
4-氯-1,2-二甲苯
分子式:
C₈H₉Cl 分子量: 140.61 g/mol 沸点: 192°C 溶点: -6 闪点: 66°C 存储条件: Ambient |
MDL编号:
MFCD00000596 CAS号: 615-60-1 EINECS: 210-438-6 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...