2-氯-1,4-二甲苯 ≥98.0% (by GC)
供应商: TCI
Warning
Synonyms: 2-氯对二甲苯
C0313-25G C0313-500G
TCIAC0313-25GEA 0 CNY
TCIAC0313-25G TCIAC0313-500G
2-氯-1,4-二甲苯 ≥98.0% (by GC)
2-氯-1,4-二甲苯
分子式: C₈H₉Cl
分子量: 140.61 g/mol
沸点: 186°C
溶点: 1.5
闪点: 57°C
MDL编号: MFCD00000564
CAS号: 95-72-7
EINECS: 202-444-2
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32089d04da5f8b7e04cd65b12d29892ca78e72ec&vid=a6f28c6a1624fe1286cc4915f40c8d6041dc2844&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %