4-吗啉基苯胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-(4-Morpholinyl)aniline

M1466-1G M1466-5G
TCIAM1466-1GEA 0 CNY
TCIAM1466-1G TCIAM1466-5G
4-吗啉基苯胺 ≥98.0% (by GC, titration analysis)
4-吗啉基苯胺
分子式: C₁₀H₁₄N₂O
溶点: 134°C
MDL编号: MFCD00006169
CAS号: 2524-67-6
EINECS: 219-760-1
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5334e61c71fdc3fe62414409adf2dfd305824e87&vid=5334e61c71fdc3fe62414409adf2dfd305824e87&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR