水杨醛肟 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-羟基苯甲醛肟

S0005-25G S0005-500G
TCIAS0005-25GEA 0 CNY
TCIAS0005-25G TCIAS0005-500G
水杨醛肟 ≥98.0% (by GC)
水杨醛肟
分子式: C₇H₇NO₂
MDL编号: MFCD00002120
CAS号: 94-67-7
EINECS: 202-353-8
Merck Index: 14,08327
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b9b6abfe8c05c98aab75298ed29401c72a53048&vid=9b9b6abfe8c05c98aab75298ed29401c72a53048&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 59.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR