N-Phenyl-p-toluidine ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Methyldiphenylamine
M1173-1G
M1173-5G
TCIAM1173-1GEA
0
CNY
TCIAM1173-1G
TCIAM1173-5G
N-Phenyl-p-toluidine ≥98.0% (by GC)
N-Phenyl-p-toluidine
分子式:
C₁₃H₁₃N 沸点: 317°C |
MDL编号:
MFCD00092921 CAS号: 620-84-8 EINECS: 210-655-6 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...