1,3-二环己基脲 ≥98.0% (by GC, titration analysis)
供应商: TCI
Synonyms:
N,N′-Dicyclohexylurea, N,N'-Dicyclohexylurea
D0441-25G
D0441-500G
TCIAD0441-25GEA
0
CNY
TCIAD0441-25G
TCIAD0441-500G
1,3-二环己基脲 ≥98.0% (by GC, titration analysis)
1,3-二环己基脲
分子式:
C₁₃H₂₄N₂O |
MDL编号:
MFCD00003829 CAS号: 2387-23-7 EINECS: 219-213-7 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 232.0 to 238.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...