4-Bromophenylurea ≥98.0% (以总氮计)

供应商: TCI

B2834-25G B2834-5G
TCIAB2834-25GEA 0 CNY
TCIAB2834-25G TCIAB2834-5G
4-Bromophenylurea ≥98.0% (以总氮计)
4-Bromophenylurea
分子式: C₇H₇BrN₂O
溶点: 227°C
MDL编号: MFCD00025428
CAS号: 1967-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d860c3ba73a36b0fcb2f8a3ef89430f8c641b48&vid=8d860c3ba73a36b0fcb2f8a3ef89430f8c641b48&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Purple to Dark purple to Dark red powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR