4-(2-羟乙基)吗啉 ≥99.0% (通过GC)
供应商: TCI
|
Warning
|
H0244-25ML
H0244-500ML
TCIAH0244-25MLEA
0
CNY
TCIAH0244-25ML
TCIAH0244-500ML
4-(2-羟乙基)吗啉 ≥99.0% (通过GC)
4-(2-羟乙基)吗啉
分子式:
C₆H₁₃NO₂ 沸点: 222°C 溶点: 2°C 闪点: 107°C 存储条件: Refrigerator |
MDL编号:
MFCD00006180 CAS号: 622-40-2 EINECS: 210-734-5 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...