3-氯-4′-氟苯丙酮 ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone

C1570-25G C1570-5G
TCIAC1570-25GEA 0 CNY
TCIAC1570-25G TCIAC1570-5G
3-氯-4′-氟苯丙酮 ≥96.0% (by GC)
3-氯-4′-氟苯丙酮
分子式: C₉H₈ClFO
分子量: 186.61 g/mol
沸点: 103°C
MDL编号: MFCD00000991
CAS号: 347-93-3
EINECS: 206-475-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a112fc50c34c76d7610b495ca8b3470ae17c403&vid=6a112fc50c34c76d7610b495ca8b3470ae17c403&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 46.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR